Learn More
CPC Scientific H-Lys-Trp-Lys-Leu-Phe-Lys-Lys-Ile-Glu-Lys-Val-Gly-Gln-Asn-Ile-Arg-Asp-Gly-Ile-Ile-Lys-Ala-Gly-Pro-Ala-Val-Ala-Val-Val-Gly-Gln-Ala-Thr-Gln-Ile-Ala-Lys-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific ATMP001B
Encompass
37-residue peptide possessing antibacterial activity against Escherichia coli, pseudomonas aeruginosa, bacillus megaterium and micrococcus luteus at micromolar concentrations. ONE-LETTER SEQUENCE: KWKLFKKIEKVGQNIRDGIIKAGPAVAVVGQATQIAK-NH2MOLECULAR FORMULA: C184H313N53O46MOLECULAR WEIGHT:4003.8STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [80451-04-3], RESEARCH AREA: AntimicrobialREFERENCES: D. Andreu et al., Proc. 20th Euro. Pept. Symp., 361 (1988 )
Catalog No. 50-210-8807