Learn More
Roche Diagnostics CEDEX CALIBRATION BEADS 20 ML
Supplier:  Roche Diagnostics 05650470001
Encompass
Cedex Calibration Beads are used to perform automated focus and brightness adjustments of the Cedex and Cedex HiRes Analyzers.
Catalog No. 50-589-342