Learn More
Apexbio Technology LLC Cedrol 77-53-2 100mg
SDP
Supplier:  Apexbio Technology LLC C6943100
Encompass_Preferred
Cedrol (CAS 77-53-2) is a small-molecule inhibitor targeting cytochrome P450 (CYP) enzymes It is designed to competitively inhibit CYP-mediated metabolic activity thereby modulating xenobiotic metabolism and related cellular processes Cedrol exerts its biological activity primarily through competitive binding to CYP enzymes In enzyme inhibition assays Cedrol demonstrates inhibitory effects with an IC50 value ranging from 15 50 M depending on CYP isoform and experimental conditions Based on these pharmacological properties Cedrol holds research potential in anti-cancer strategies management of oxidative stress and inflammatory responses neuropathic pain models antibacterial applications anxiety reduction and the prevention of hair loss
Catalog No. 50-000-90156