Learn More
Medchemexpress LLC Ceefourin 1 | 315702-40-0 | MFCD02329957 | 294.42 g·mol⁻¹ | C11H10N4S3 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY101453100MG
Encompass_Preferred
Ceefourin 1 is a potent, selective inhibitor of multidrug resistance protein 4 (MRP4/ABCC4) supplied for research and analytical applications. It is used in transporter assays and cell-based studies to probe MRP4-mediated efflux and as a chemosensitizer probe in preclinical research. The compound has formula C11H10N4S3 and a molecular weight of 294.42 g·mol⁻¹.
  • Selective MRP4 inhibitor for transporter studies.
  • Useful as an analytical standard for assay development and validation.
  • Reported low cellular toxicity in standard assays.
  • Soluble in DMSO, suitable for in vitro use.
  • Available as a small-quantity powder for research applications.
Catalog No. 50-002-48838