Learn More
AnaSpec CEF1, Influenza Matrix Protein M1 (58 - 66), 1mg
Supplier:  AnaSpec AS28310
Encompass
CEF1, Influenza Matrix Protein M1 (58-66)[GILGFVFTL] ; MW 966.2; (1 mg) HLA A2-restricted epitope from influenza matrix protein (58-66).
Catalog No. NC0893836