Learn More
AnaSpec CEF20, Cytomegalovirus, CMV pp65 (495-503)[NLVPMVATV] | MW 943.2 | (1 mg) HLA-A*0201-restricted epitope from Cytomegalovirus pp65 (495-503).
Supplier:  AnaSpec AS28328
Encompass
CEF20, Cytomegalovirus, CMV pp65 (495-503)[NLVPMVATV] ; MW 943.2; (1 mg) HLA-A*0201-restricted epitope from Cytomegalovirus pp65 (495-503).
Catalog No. NC0892283