Learn More
Apexbio Technology LLC Cefaclor 53994-73-3 5g
SDP
Supplier:  Apexbio Technology LLC A55595000
Encompass_Preferred
Cefaclor (CAS 53994-73-3) is a semi-synthetic cephalosporin antibiotic targeting penicillin-binding proteins (PBPs) It inhibits these enzymes thereby disrupting the bacterial cell wall biosynthesis pathway Cefaclor exerts its biological activity primarily through inhibition of the transpeptidation step in peptidoglycan synthesis In in vitro studies Cefaclor demonstrates bactericidal activity with minimal inhibitory concentration (MIC) values generally ranging from 1 g/mL to 16 g/mL depending on the bacterial species tested Based on these pharmacological properties Cefaclor holds research potential in the fields of antibiotic resistance profiling bacterial susceptibility testing and studies involving gram-positive and gram-negative bacterial infections
Catalog No. 50-000-93732