Learn More
Medchemexpress LLC Cefaclor | 53994-73-3 | 98.5% | 1 G
SDP
Supplier:  Medchemexpress LLC HYB01981G
Encompass_Preferred
Cefaclor is a well-absorbed orally active cephalosporin antibiotic. It specifically binds to penicillin-binding protein 3 (PBP3) and can be used for research on depression and various bacterial infections, including respiratory tract infections, bacterial bronchitis, pharyngitis, and skin infections. This product is intended for research and analytical applications only and has not been fully validated for medical use.
  • Well-absorbed orally active cephalosporin antibiotic
  • Specifically binds to penicillin-binding protein 3 (PBP3)
  • Used for research on depression
  • Effective against various bacterial infections including respiratory tract infections, bacterial bronchitis, pharyngitis, and skin infections
  • Provided as a light yellow to yellow solid
Catalog No. 50-004-05703