Learn More
Medchemexpress LLC Cefaclor | 53994-73-3 | 98.5% | 5 G
SDP
Supplier:  Medchemexpress LLC HYB01985G
Encompass_Preferred
Cefaclor is a well-absorbed orally active cephalosporin antibiotic. It specifically binds to penicillin-binding protein 3 (PBP3) and is used for research into depression and various bacterial infections, including respiratory tract infections, bacterial bronchitis, pharyngitis, and skin infections.
  • Well-absorbed orally active cephalosporin antibiotic
  • Specifically binds to penicillin-binding protein 3 (PBP3)
  • Used for research of depression and various bacterial infections
  • Shows significant antibacterial activity against Gram-positive and Gram-negative isolates
Catalog No. 50-004-05931