Learn More
TARGETMOL CHEMICALS INC Cefaclor monohydrate 200MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T1495200MG
Encompass_Preferred
Also available in 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Cefaclor monohydrate (Cefaclorum) is a beta-lactam, second-generation cephalosporin antibiotic with bactericidal activity. Purity 99.48%
Catalog No. 50-287-6405