Learn More
Selleck Chemical LLC Cefadroxil E4906-100MG
SDP
Supplier:  Selleck Chemical LLC E4906100MG
Encompass_Preferred
Cefadroxil(BL-S 578) is a first-generation cephalosporin antibiotic with broad spectrum of anti-bacterial activity It can be used in the management of infections in the respiratory tract urinary tract skin and soft tissues and bones and joints
Catalog No. 50-000-43358