Learn More
Cayman Chemical Cefamndole Nafate 1g
SDP
Supplier:  Cayman Chemical 239881 G
Encompass_Preferred
A prodrug from of cefamandole; inhibits the growth of E. cloacae, E. coli, K. pneumoniae, P. mirabilis, P. vulgaris, P. morganii, S. aureus, and S. pyogenes in vitro (MICs = 0.1-50 μg/ml); protects against E. cloacae, E. coli, K. pneumoniae, P. mirabilis, P. vulgaris, P. morganii, S. aureus, and S. pyogenes infection in mice (ED50s = 0.3-334 mg/kg); protects against tobramycin-induced nephrotoxicity at 500 mg/kg
Catalog No. 50-293-3191