Learn More
Cayman Chemical CefamndolesodIum salt 50mg
SDP
Supplier:  Cayman Chemical 1813850 MG
Encompass_Preferred
A cephalosporin antibiotic that is effective against E. coli (MIC values range from 0.25-2 mg/L depending on strain) as well as H. influenza and S. aureus
Catalog No. 50-296-4888