Learn More
Research Products International Corp Cefazolin Sodium Salt, 500 MG
SDP
Supplier:  Research Products International Corp C545000.5
Encompass_Preferred
Cefazolin Sodium Salt is a first-generation cephalosporin antibiotic. It inhibits bacterial cell wall synthesis by binding to penicillin-binding proteins (PBPs), enzymes involved in the final stages of peptidoglycan synthesis. This interference leads to structural instability and eventual cell lysis. 
Effective against a broad spectrum of bacteria, particularly Gram-positive organisms.  Can be used in microbiology research and studies focusing on antibacterial agents. Its inclusion in culture media can help selectively isolate bacteria that are susceptible to this antibiotic.
Catalog No. 50-197-7758