Learn More
Selleck Chemical LLC Cefdinir 100mg 91832-40-5 FK 482, PD 134393, CI-983
SDP
Supplier:  Selleck Chemical LLC S1605100MG
Encompass_Preferred
Cefdinir (FK 482, PD 134393, CI-983) is an oral cephalosporin antibiotic, used to treat bacterial infections in many different parts of the body. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-678-8