Learn More
Medchemexpress LLC Cefditoren Pivoxil | 117467-28-4 | MFCD00926593 | 99.3% | 620.72 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY17452A50MG
Encompass_Preferred
Cefditoren Pivoxil (ME 1207) is a broad-spectrum, third-generation, oral cephalosporin antibacterial. It shows enhanced stability against many common β lactamases and is active against both Gram-negative and Gram-positive organisms. This compound can be used in research for infectious diseases such as acute exacerbations of chronic bronchitis, community-acquired pneumonia (CAP), streptococcal pharyngitis/tonsillitis, or uncomplicated skin and skin structure infections.
  • Broad-spectrum, third-generation, oral cephalosporin antibacterial.
  • Enhanced stability against many common β lactamases.
  • Activity against Gram-negative and Gram-positive organisms.
  • Useful in research for various infectious diseases.
Catalog No. 50-003-86592