Learn More
Medchemexpress LLC Cefditoren (sodium) | 104146-53-4 | 99.7% | 528.56 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY17452
Encompass_Preferred
Cefditoren sodium (ME 1206) is a broad-spectrum, third-generation, oral cephalosporin antibacterial. It has enhanced stability against many common β lactamases and is active against Gram-negative and Gram-positive organisms. It is used in the research of infectious diseases such as acute exacerbations of chronic bronchitis, community-acquired pneumonia (CAP), streptococcal pharyngitis/tonsillitis, or uncomplicated skin and skin structure infections.
  • Broad-spectrum, third-generation, oral cephalosporin antibacterial
  • Enhanced stability against common β lactamases
  • Active against Gram-negative and Gram-positive organisms
  • Suitable for research of infectious diseases
Catalog No. 50-003-84131