Learn More
Medchemexpress LLC Cefditoren (sodium) | 104146-53-4 | 99.7% | 528.56 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY174525MG
Encompass_Preferred
Cefditoren sodium (ME 1206) is a broad-spectrum, third-generation, oral cephalosporin antibacterial with enhanced stability against many common β lactamases. It has activity against Gram-negative and Gram-positive organisms. It can be used in research for infectious diseases such as acute exacerbations of chronic bronchitis, community-acquired pneumonia (CAP), streptococcal pharyngitis/tonsillitis, or uncomplicated skin and skin structure infections.
  • Broad-spectrum, third-generation, oral cephalosporin antibacterial
  • Enhanced stability against many common β lactamases
  • Active against Gram-negative and Gram-positive organisms
  • Effective for research on infectious diseases
  • Shows good activity against common bacteria like Streptococcus pneumoniae, S. pyogenes, and Staphylococcus aureus
Catalog No. 50-003-86679