Learn More
Cayman Chemical CefdItren PIvoxIl 5mg
SDP
Supplier:  Cayman Chemical 303155 MG
Encompass_Preferred
A prodrug form of cefditoren; effective against systemic S. aureus, E. coli, K. pneumoniae, P. mirabilis, or S. marcescens infections in mice (ED50s = 10, 2.5, 11, 4.4, and 11 mg/kg, respectively)
Catalog No. 50-298-8566