Learn More
Apexbio Technology LLC Cefepime Dihydrochloride Monohydrate(Synonyms: Cefepime hydrochloride, Cefepime HCl, Maxipime, Cefepime hydrochloride hydrate), 5g, CAS: 123171-59-5.
SDP
Supplier:  Apexbio Technology LLC B35195000
Encompass_Preferred
Cefepime Dihydrochloride Monohydrate (CAS 123171-59-5) is a cephalosporin antibiotic that acts by inhibiting bacterial cell wall synthesis disrupting the structural integrity and causing cell death Experimental data indicate antimicrobial activity against both Gram-positive and Gram-negative bacteria This compound is utilized in clinical investigations targeting complex urinary tract infections severe pneumonia and multidrug-resistant bacterial infections
Catalog No. 50-000-60434