Learn More
Selleck Chemical LLC Cefepime E7089-50MG
SDP
Supplier:  Selleck Chemical LLC E708950MG
Encompass_Preferred
Cefepime (BMY-28142) is a broad-spectrum and cross the blood-brain barrier cephalosporin Cefepime shows antibacterial effects against both Gram-positive and Gram-negative aerobic bacteria Cefepime induces neurotoxicity
Catalog No. 50-000-41321