Learn More
Medchemexpress LLC Cefepime-d8 sulfate | 131857-33-5 | 99.2% | C19H18D8N6O9S3 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY143831S1MG
Encompass_Preferred
Cefepime-d8 sulfate is a deuterium-labeled form of Cefepime sulfate, intended for research purposes. It is useful for understanding drug pharmacokinetics and metabolic profiles.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-55898