Learn More
Medchemexpress LLC Cefepime Dihydrochloride Monohydrate | 123171-59-5 | 99.91% | 5 G
SDP
Supplier:  Medchemexpress LLC HYB06165G
Encompass_Preferred
Cefepime Dihydrochloride Monohydrate is a broad-spectrum cephalosporin known for its enhanced coverage against both Gram-positive and Gram-negative bacteria. As an extended-spectrum parenteral cephalosporin antibiotic, it exhibits activity in vitro against a wide range of gram-positive and gram-negative aerobic bacteria. Notably, Cefepime demonstrates a reduced tendency to induce beta-lactamases compared to other beta-lactam antibiotics. Its pharmacokinetic profile is similar to other renally eliminated cephalosporins, with a half-life of approximately 2 hours. This compound has shown clinical effectiveness in treating various infections and is generally well tolerated.
  • Broad-spectrum cephalosporin with enhanced coverage
  • Active against gram-positive and gram-negative aerobic bacteria
  • Decreased propensity to induce beta-lactamases
  • Demonstrated clinical efficacy against various infections
  • Generally well tolerated
Catalog No. 50-004-08319