Learn More
Medchemexpress LLC Cefetamet | 65052-63-3 | 99.1% | C14H15N5O5S2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYA0111100MG
Encompass_Preferred
Cefetamet is a cephalosporin antibiotic and the active metabolite of Cefetamet pivoxil. It binds to bacterial penicillin-binding protein (PBP), demonstrating significant activity against Gram-negative bacteria such as Enterobacteriaceae, Neisseria species, and Haemophilus influenzae, as well as Gram-positive bacteria like Streptococcus. Cefetamet kills and lyses Treponema pallidum and is utilized in research for various infections including respiratory tract, urinary tract, ear, nose, and throat infections, and syphilis.
  • Cephalosporin antibiotic
  • Active against Gram-negative and Gram-positive bacteria
  • Binds to bacterial penicillin-binding protein (PBP)
  • Kills and lyses Treponema pallidum
  • Useful in research for a variety of infections
Catalog No. 50-004-32558