Learn More
Apexbio Technology LLC Cefetamet pivoxil hydrochloride 111696-23-2 25mg
SDP
Supplier:  Apexbio Technology LLC C648525
Encompass_Preferred
Cefetamet pivoxil hydrochloride (CAS 111696-23-2) is a third-generation cephalosporin antibiotic targeting penicillin-binding proteins It is designed to inhibit these enzymes thereby disrupting bacterial cell wall biosynthesis in Gram-positive and Gram-negative bacteria Cefetamet pivoxil hydrochloride exerts its biological activity primarily through inhibition of peptidoglycan synthesis In in vitro antibacterial assays cefetamet pivoxil hydrochloride demonstrates inhibitory activity with IC50 values within the micromolar range in clinical isolates associated with respiratory and urinary infections Based on these pharmacological properties cefetamet pivoxil hydrochloride holds research potential in studies of bacterial enzyme inhibition antibiotic resistance mechanisms and antimicrobial susceptibility profiling
Catalog No. 50-000-90087