Learn More
Apexbio Technology LLC Cefiderocol 1225208-94-5 10mg
SDP
Supplier:  Apexbio Technology LLC BA100410
Encompass_Preferred
Cefiderocol (CAS 1225208-94-5) is a siderophore cephalosporin antibiotic with potent and broad-spectrum activity against aerobic Gram-negative bacteria It utilizes a Trojan horse strategy by exploiting bacterial iron transport systems to facilitate cell entry enabling inhibition of cell wall synthesis through binding to penicillin-binding proteins This compound is used in research to study mechanisms of antimicrobial resistance and to evaluate novel therapeutic approaches against multidrug-resistant Gram-negative pathogens
Catalog No. 50-000-89439