Learn More
Medchemexpress LLC HY-17628 25mg , Cefiderocol CAS:1225208-94-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1762825MG
Encompass_Preferred
HY-17628 25mg Cefiderocol CAS: 1225208-94-5 Cefiderocol (S-649266) is a siderophore cephalosporin which has a potent activity against a broad range of aerobic Gram-negative bacterial species with MIC50s of 2 μg/mL or less. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9267