Learn More
Selleck Chemical LLC Cefiderocol-E1792-1MG
SDP
Supplier:  Selleck Chemical LLC E17921MG
Encompass_Preferred
Cefiderocol (S-649266) is a siderophore cephalosporin which has a potent activity against a broad range of aerobic Gram-negative bacterial species with MIC50s of 2 g/mL or less
Catalog No. 50-313-8880