Learn More
Medchemexpress LLC Cefixime trihydrate | 125110-14-7 | 99.8% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB1381A100MG
Encompass_Preferred
Cefixime trihydrate is an antibiotic and a third-generation cephalosporin, useful for the treatment of a number of bacterial infections. It is for research use only and not for patient use.
  • Shows great antibacterial activity against clinical isolates of *Salmonella typhi*.
  • Active against β-lactamase producing Amoxicillin-resistant strains.
  • Active against *Enterobacteriaceae*, such as *Haemophilus influenzae*, and *Neisseria gonorrhoeae*.
  • Available in various sizes.
  • Purity of 99.8%.
  • White to off-white solid appearance.
Catalog No. 50-004-12119