Learn More
Medchemexpress LLC Cefmenoxime hydrochloride | 75738-58-8 | 99.29% | 500 MG
SDP
Supplier:  Medchemexpress LLC HYB0875500MG
Encompass_Preferred
Cefmenoxime (SCE-1365) hydrochloride is a new semisynthetic cephalosporin antibiotic that demonstrates antibacterial activity against a wide variety of gram-positive and gram-negative bacteria.
  • Inhibits at least 90% of tested strains (MIC90) ranging from 0.06 to 8 μg/mL for Enterobacteriaceae.
  • MIC90 values for gram-positive cocci are 0.015 and ≤0.008 μg/mL for Streptococcus pneumoniae and Streptococcus pyogenes, respectively, and 2 μg/mL for S. aureus.
  • Exhibits activity against Haemophilus influenzae, Neisseria gonorrhoeae, and Neisseria meningitidis with MIC90 values from ≤0.008 to 0.25 μg/mL.
  • Improved the survival rate of mice infected with lung bacteria in in vivo studies.
Catalog No. 50-004-08854