Learn More
Medchemexpress LLC Cefmenoxime hydrochloride | 75738-58-8 | 99.3% | 5 G
SDP
Supplier:  Medchemexpress LLC HYB08755G
Encompass_Preferred
Cefmenoxime hydrochloride is a new semisynthetic cephalosporin antibiotic. It demonstrates antibacterial activity against a broad range of gram-positive and gram-negative bacteria.
  • Acts as a β-lactam antibiotic.
  • Exhibits broad-spectrum activity against various bacteria.
  • Inhibits at least 90% of tested Enterobacteriaceae strains (MIC90 0.06 to 8 μg/mL) in vitro.
  • Effective against Streptococcus pneumoniae (MIC90 0.015 μg/mL), Streptococcus pyogenes (MIC90 ≤0.008 μg/mL), and S. aureus (MIC90 2 μg/mL) in vitro.
  • Shows activity against Haemophilus influenzae, Neisseria gonorrhoeae, and Neisseria meningitidis (MIC90 ≤0.008 to 0.25 μg/mL) in vitro.
  • Improves survival rate in mice infected with lung bacteria (40 mg/kg, daily for 7 days, 60% mortality reduction) in vivo.
Catalog No. 50-004-08076