Learn More
Selleck Chemical LLC Cefonicid sodium S3672-25mg
SDP
Supplier:  Selleck Chemical LLC S367225MG
Encompass_Preferred
Cefonicid sodium (Monocid) is a cephalosporin antibiotic It interferes with cell wall biosynthesis in bacteria leading to lysis of the infectious organism
Catalog No. 50-310-8920