Learn More
Medchemexpress LLC Cefoperazone-d5 | 2410425-70-4 | 98.6% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0210S1MG
Encompass_Preferred
Cefoperazone-d5 is a deuterium-labeled form of Cefoperazone, a semisynthetic cephalosporin known for its broad-spectrum antibacterial activity. This compound is intended for research use only, primarily as a tracer or an internal standard in quantitative analytical methods.
  • Deuterium labeled cefoperazone
  • Semisynthetic cephalosporin with broad-spectrum antibacterial activity
  • Functions as a tracer in research
  • Used as an internal standard for quantitative analysis (NMR, GC-MS, LC-MS)
  • Incorporates stable heavy isotopes for drug development quantitation
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
  • Targets β-lactam
Catalog No. 50-004-05636