Learn More
Alkali Scientific Cefoperazone, Sodium Salt, 5 Grams
SDP
Supplier:  Alkali Scientific EFO001CP
Encompass_Preferred
Cefoperazone sodium salt is a broad-spectrum cephalosporin antibiotic supplied in a 5 gram laboratory-grade format for research and microbiological applications. It is effective against a wide range of Gram-negative and certain Gram-positive bacteria by inhibiting bacterial cell wall synthesis. This compound is commonly used in selective media formulations, antimicrobial susceptibility testing, and microbiological research where controlled bacterial inhibition is required. The sodium salt form provides improved solubility in aqueous solutions, making it suitable for laboratory preparation and experimental workflows. It is widely utilized in pharmaceutical, clinical, and research laboratories for antimicrobial studies and culture media development.
Catalog No. 50-006-60365