Learn More
Medchemexpress LLC Cefotaxime sodium | 64485-93-4 | 99.1% | C16H16N5NaO7S2 | 1 G
SDP
Supplier:  Medchemexpress LLC HYA00881G
Encompass_Preferred
Cefotaxime sodium is a β-lactamase stable cephalosporin and a third-generation cephalosporin antibiotic. It demonstrates broad-spectrum antibiotic activity against numerous Gram-positive and Gram-negative bacteria.
  • Beta-lactamase stable cephalosporin
  • Third-generation cephalosporin antibiotic
  • Broad-spectrum antibiotic activity against Gram-positive and Gram-negative bacteria
  • Exhibits an MIC of 0.0625 mg/L for V. vulnificus CMCP6
  • Effective in clearing V. vulnificus in vivo when combined with ciprofloxacin
Catalog No. 50-004-32131