Learn More
Medchemexpress LLC Cefotaxime sodium | 64485-93-4 | 99.1% | C16H16N5NaO7S2 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYA0088A
Encompass_Preferred
Cefotaxime sodium is a beta-lactamase stable cephalosporin and a third-generation cephalosporin antibiotic. It demonstrates broad-spectrum antibiotic activity against numerous Gram-positive and Gram-negative bacteria. This product is for research use only.
  • Broad-spectrum antibiotic activity
  • Effective against Gram-positive and Gram-negative bacteria
  • Beta-lactamase stable
  • Exhibits potent antimicrobial activity against V. vulnificus
Catalog No. 50-003-58580