Learn More
Enzo Life Sciences Cefotaxime . sodium salt (1g). CAS: 64485-93-4
SDP
Supplier:  Enzo Life Sciences ALX380271G001
Encompass_Preferred
Cephalosporin antibiotic. Inhibits bacterial cell wall synthesis. Effective against Gram-negative and Gram-positive bacteria. Resistant against β-lactamase. Purity: ≥95% (HPLC). Appearance: White to light yellow powder. Solubility: Soluble in water (>50mg/ml). Long Term Storage: -20°C.
Catalog No. 50-200-9003