Learn More
TOKU-E CEFOTAXIME SOLN 10X1ML
Supplier:  TOKU-E C29310X1ML
Encompass_Preferred
Antibiotic, ReadyMade™, sterile-filtered, Cefotaxime Sodium, 100 mg/ml in water. Useful in microbiology and plant biology.
Catalog No. NC2402954