Learn More
Cayman Chemical CefotaxImesodIum salt 1g
SDP
Supplier:  Cayman Chemical 160401 G
Encompass_Preferred
A cephalosporin antibiotic with potent in vitro activity against streptococcal species, including S. aureus (IC50 = 2 UG/ml) and penicillin-resistant S. pneumonia and also effective against a wide range of other β-lactamase-producing bacterial species (MIC90s = 0.01 - 130 UG/ml)
Catalog No. 50-296-8984