Learn More
Selleck Chemical LLC Cefotetan disodium salt E2881-25MG
SDP
Supplier:  Selleck Chemical LLC E288125MG
Encompass_Preferred
Cefotetan disodium salt is a semisynthetic cephamycin antibiotic that exerts its bactericidal effects by inhibition of cell-wall synthesis It showed in vitro efficacy against ESBL-producing E coli bloodstream isolates with MIC50 of 0 25 mg/L
Catalog No. 50-000-39748