Learn More
TARGETMOL CHEMICALS INC Cefotiam Hexetil Hydrochlor
SDP
Supplier:  TARGETMOL CHEMICALS INC T502410MG
Encompass_Preferred
Also available in 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Cefotiam Hexetil Hydrochloride is a third-generation, semi-synthetic, beta-lactam cephalosporin antibiotic with antibacterial activity. Purity 100%
Catalog No. 50-287-7334