Learn More
Cayman Chemical CefotIam hexetIlhydrochLRI 5mg
SDP
Supplier:  Cayman Chemical 395385 MG
Encompass_Preferred
A pan-selenoprotein inhibitor and prodrug form of cefotiam; inhibits GPX1, GPX4, and TrxR1 in cell-free assays from 1-100 µM
Catalog No. 50-291-9159