Learn More
Selleck Chemical LLC Cefotiam hydrochloride S5005-25mg
SDP
Supplier:  Selleck Chemical LLC S500525MG
Encompass_Preferred
Cefotiam hydrochloride (Halospor Pansporin Pansporine) is the hydrochloride salt form of cefotiam and has a broad spectrum of activity against both gram-positive and gram-negative microorganisms
Catalog No. 50-311-1736