Learn More
Cayman Chemical Cefotiam hydrochloride hydra
SDP
Supplier:  Cayman Chemical 32871500 MG
Encompass_Preferred
A broad-spectrum cephalosporin antibiotic; inhibits the growth of S. aureus (MIC50 = 0.26 UG/ml), as well as P. aeruginosa, E. coli, and H. influenzae (MIC50s = 1, 0.06, and 0.01 UG/ml, respectively); increases survival in mouse models of infection with strains of S. aureus (ED50s = 3.5 and 8.3 mg/kg) and E. coli (ED50s = 0.5-25 mg/kg)
Catalog No. 50-294-8326