Learn More
Medchemexpress LLC Cefovecin sodium | 141195-77-9 | 99.8% | 475.47 | C17H18N5NaO6S2 | 10 MG

Supplier: Medchemexpress LLC HY11768510MG
Cefovecin sodium is the sodium salt of cefovecin, a third-generation cephalosporin antibiotic supplied as a high-purity research reagent. It interferes with bacterial cell wall synthesis and is commonly used in vitro to study antibacterial activity and assay development.
- High purity: 99.79% (reported by supplier).
- Molecular formula C17H18N5NaO6S2.
- Molecular weight 475.47 g/mol.
- Supplied as a solid suitable for laboratory use.
- Available in small research pack sizes, e.g., 10 MG, for assay-scale experiments.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.