Learn More
Medchemexpress LLC Cefoxitin | 35607-66-0 | 99.92% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB182510MM/1ML
Encompass_Preferred
Cefoxitin is a cephalosporin-class antibiotic that is highly stable against β-Lactamase. It has a broad spectrum of antibacterial activity, effective against both Gram-negative and Gram-positive bacteria. This product is for research use only.
  • Cephalosporin-class antibiotic
  • Highly stable against β-Lactamase
  • Broad spectrum of antibacterial activity
  • Effective against Gram-negative bacteria
  • Effective against Gram-positive bacteria
  • Inhibits mouse OAT3 expressed in CHO cells
  • Effectively kills *B. burgdorferi* in mice
Catalog No. 50-004-12801