Learn More
Selleck Chemical LLC Cefozopran hydrochloride
SDP
Supplier:  Selleck Chemical LLC S563825MG
Encompass_Preferred
Cefozopran Hydrochloride is the hydrochloride salt form of cefozopran a semi-synthetic broad-spectrum fourth-generation cephalosporin with antibacterial activity
Catalog No. 50-000-03163