Learn More
Medchemexpress LLC Cefpirome sulfate | 98753-19-6 | 99.5% | 5 G
Supplier: Medchemexpress LLC HYB18245G
Cefpirome sulfate is a cephalosporin antibiotic that can cross cell membranes and the blood-brain barrier. It binds to penicillin-binding proteins with high affinity, inhibiting bacterial cell wall synthesis. Cefpirome sulfate exhibits bactericidal and growth-inhibitory activities against:
- Gram-negative bacteria
- Gram-positive bacteria
- Susceptible anaerobic bacteria (including some β-lactamase-producing strains)