Learn More
Cayman Chemical CefpodoxIme-d3 1mg
SDP
Supplier:  Cayman Chemical 254121 MG
Encompass_Preferred
An internal standard for the quantification of cefpodoxime by GC- or LC-MS
Catalog No. 50-293-3434