Learn More
Cayman Chemical CefpodoxIme-d3 500ug
SDP
Supplier:  Cayman Chemical 25412500 UG
Encompass_Preferred
An internal standard for the quantification of cefpodoxime by GC- or LC-MS
Catalog No. 50-293-3100